Anti-NANOS1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50-1:200
Type:
Antigen: NANOS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-418
- 77721-417
- Antigen Symbol:
- NANOS1
- NANOS1
- Antigen Synonyms:
- Nanos Homologue 1 (NANOS1)|EC_Rep1a|NANOS 1|NANOS1|Nanos homolog 1 (Drosophila)|EC Rep1a|Rep1a|Rep 1a|NOS-1|OTTHUMP00000046853|Nos1|Nos 1|NANO1_HUMAN|Nanos homolog 1
- Nanos Homologue 1 (NANOS1)|EC_Rep1a|NANOS 1|NANOS1|Nanos homolog 1 (Drosophila)|EC Rep1a|Rep1a|Rep 1a|NOS-1|OTTHUMP00000046853|Nos1|Nos 1|NANO1_HUMAN|Nanos homolog 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WY41
- Q8WY41
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Nanos homolog 1 protein (1-292AA)
- Recombinant Human Nanos homolog 1 protein (1-292AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together