Anti-CXorf40A Rabbit Polyclonal Antibody
New Product
Type:
Antigen: CXorf40A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-260
- 77721-259
- Antigen Symbol:
- CXorf40A
- CXorf40A
- Antigen Synonyms:
- CXorf40A|Endothelial-overexpressed lipopolysaccharide-associated factor 1|EOLA1Protein CXorf40A|CXorf40
- CXorf40A|Endothelial-overexpressed lipopolysaccharide-associated factor 1|EOLA1Protein CXorf40A|CXorf40
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE69
- Q8TE69
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein CXorf40A protein (21-158AA)
- Recombinant Human Protein CXorf40A protein (21-158AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together