Anti-ADAMTS15 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ADAMTS15
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-254
- 77721-253
- Antigen Symbol:
- ADAMTS15
- ADAMTS15
- Antigen Synonyms:
- ADAMTS-15|ATS15_HUMAN|MGC126403|ADAM metallopeptidase with thrombospondin type 1 motif 15|ADAM TS 15|ADAMTS15|ADAM-TS 15|ADAM-TS15|ADAM TS15|A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15 preproprotein|A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15|A disintegrin like and metalloprotease with thrombospondin type 1 motif 15 preproprotein|A disintegrin like and metalloprotease with thrombospondin type 1 motif 15|ADAMTS 15|A disintegrin and metalloproteinase with thrombospondin motifs 15
- ADAMTS-15|ATS15_HUMAN|MGC126403|ADAM metallopeptidase with thrombospondin type 1 motif 15|ADAM TS 15|ADAMTS15|ADAM-TS 15|ADAM-TS15|ADAM TS15|A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15 preproprotein|A disintegrin like and metalloprotease reprolysin type with thrombospondin type 1 motif 15|A disintegrin like and metalloprotease with thrombospondin type 1 motif 15 preproprotein|A disintegrin like and metalloprotease with thrombospondin type 1 motif 15|ADAMTS 15|A disintegrin and metalloproteinase with thrombospondin motifs 15
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TE58
- Q8TE58
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 15 protein (801-913AA)
- Recombinant Human A disintegrin and metalloproteinase with thrombospondin motifs 15 protein (801-913AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together