Anti-KDM2B Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: KDM2B
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77721-163
- 77721-162
- Antigen Symbol:
- KDM2B
- KDM2B
- Antigen Synonyms:
- Lysine-specific demethylase 2B|PCCX2|F-box protein FBL10|Protein JEMMA|F box and leucine rich repeat protein 10|Jumonji C domain containing histone demethylase 1B|F box protein FBL10|Lysine (K) specific demethylase 2B|JEMMA (Jumonji domain EMSY interactor methyltransferase motif) protein|Protein containing CXXC domain 2|JHDM1B|Jumonji domain-containing EMSY-interactor methyltransferase motif protein|F-box and leucine-rich repeat protein 10|CXXC-type zinc finger protein 2|JmjC domain-containing histone demethylation protein 1B|CXXC2|Fbl10|kdm2b|KDM2B_HUMAN|F-box/LRR-repeat protein 10|[Histone-H3]-lysine-36 demethylase 1B|FBXL10|Protein-containing CXXC domain 2
- Lysine-specific demethylase 2B|PCCX2|F-box protein FBL10|Protein JEMMA|F box and leucine rich repeat protein 10|Jumonji C domain containing histone demethylase 1B|F box protein FBL10|Lysine (K) specific demethylase 2B|JEMMA (Jumonji domain EMSY interactor methyltransferase motif) protein|Protein containing CXXC domain 2|JHDM1B|Jumonji domain-containing EMSY-interactor methyltransferase motif protein|F-box and leucine-rich repeat protein 10|CXXC-type zinc finger protein 2|JmjC domain-containing histone demethylation protein 1B|CXXC2|Fbl10|kdm2b|KDM2B_HUMAN|F-box/LRR-repeat protein 10|[Histone-H3]-lysine-36 demethylase 1B|FBXL10|Protein-containing CXXC domain 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NHM5
- Q8NHM5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lysine-specific demethylase 2B protein (1274-1336AA)
- Recombinant Human Lysine-specific demethylase 2B protein (1274-1336AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together