Anti-BBS1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: BBS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77690-236
- 77721-144
- Antigen Symbol:
- BBS1
- BBS1
- Antigen Synonyms:
- BBS2L2|AI451249|Bardet-Biedl syndrome 1|D19Ertd609e|Bardet-Biedl syndrome 1 homolog|BBS2-like protein 2|BBS1|BBS1_HUMAN|Bardet-Biedl syndrome 1 protein
- BBS2L2|AI451249|Bardet-Biedl syndrome 1|D19Ertd609e|Bardet-Biedl syndrome 1 homolog|BBS2-like protein 2|BBS1|BBS1_HUMAN|Bardet-Biedl syndrome 1 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFJ9
- Q8NFJ9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Bardet-Biedl syndrome 1 protein (217-417AA)
- Recombinant Human Bardet-Biedl syndrome 1 protein (217-417AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together