Anti-NUP35 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUP35
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-140
- 77721-141
- Antigen Symbol:
- NUP35
- NUP35
- Antigen Synonyms:
- NUP53_HUMAN|NP44|NUP53|Nucleoporin 35kDa|NP 44|NUP35|35 kDa nucleoporin|Nuclear pore complex protein 53kDa|MP-44|Nucleoporin 53kDa|Nuclear pore protein NUP53|Nuclear pore complex protein Nup53|Nucleoporin Nup 35|Nup 53|Nuclear pore complex protein Nup 53|Mitotic phosphoprotein 44|Nucleoporin Nup35|NUP 35|Nuclear pore protein NUP 53|MP 44|Nucleoporin NUP 53|Nucleoporin NUP53
- NUP53_HUMAN|NP44|NUP53|Nucleoporin 35kDa|NP 44|NUP35|35 kDa nucleoporin|Nuclear pore complex protein 53kDa|MP-44|Nucleoporin 53kDa|Nuclear pore protein NUP53|Nuclear pore complex protein Nup53|Nucleoporin Nup 35|Nup 53|Nuclear pore complex protein Nup 53|Mitotic phosphoprotein 44|Nucleoporin Nup35|NUP 35|Nuclear pore protein NUP 53|MP 44|Nucleoporin NUP 53|Nucleoporin NUP53
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NFH5
- Q8NFH5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Nucleoporin NUP53 protein (6-305AA)
- Recombinant Human Nucleoporin NUP53 protein (6-305AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together