Anti-WDR19 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WDR19
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-133
- 77721-134
- Antigen Symbol:
- WDR19
- WDR19
- Antigen Synonyms:
- WD repeat-containing protein 19|DYF 2|Wdr19|FLJ23127|PWDMP|Intraflagellar transport 144 homolog|KIAA1638|IFT144|WD repeat containing protein 19|DYF2|WDR19_HUMAN|ORF26|WD repeat domain 19|WD repeat membrane protein PWDMP|Oseg6
- WD repeat-containing protein 19|DYF 2|Wdr19|FLJ23127|PWDMP|Intraflagellar transport 144 homolog|KIAA1638|IFT144|WD repeat containing protein 19|DYF2|WDR19_HUMAN|ORF26|WD repeat domain 19|WD repeat membrane protein PWDMP|Oseg6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NEZ3
- Q8NEZ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human WD repeat-containing protein 19 protein (45-226AA)
- Recombinant Human WD repeat-containing protein 19 protein (45-226AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together