Anti-VGLL2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VGLL2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-054
- 77721-055
- Antigen Symbol:
- VGLL2
- VGLL2
- Antigen Synonyms:
- VITO1|Transcription cofactor vestigial-like protein 2|Protein VITO1|VGLL2|Vgl-2
- VITO1|Transcription cofactor vestigial-like protein 2|Protein VITO1|VGLL2|Vgl-2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N8G2
- Q8N8G2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transcription cofactor vestigial-like protein 2 protein (1-143AA)
- Recombinant Human Transcription cofactor vestigial-like protein 2 protein (1-143AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together