Anti-HORMAD2 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HORMAD2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-033
- 77721-034
- Antigen Symbol:
- HORMAD2
- HORMAD2
- Antigen Synonyms:
- RP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2
- RP23-329P17.4|MGC26710|HORM2_HUMAN|HORMA domain-containing protein 2|HORMA domain containing 2|CT46.2|4930529M09Rik|Hormad2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N7B1
- Q8N7B1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human HORMA domain-containing protein 2 protein (14-307AA)
- Recombinant Human HORMA domain-containing protein 2 protein (14-307AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together