Anti-LZTR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: LZTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77721-001
- 77721-000
- Antigen Symbol:
- LZTR1
- LZTR1
- Antigen Synonyms:
- Leucine-zipper-like transcriptional regulator 1|Leucine zipper like transcription regulator 1|TCFL2|LZTR1_HUMAN|Leucine zipper like regulator 1|MGC21205|Lztr1|LZTR-1
- Leucine-zipper-like transcriptional regulator 1|Leucine zipper like transcription regulator 1|TCFL2|LZTR1_HUMAN|Leucine zipper like regulator 1|MGC21205|Lztr1|LZTR-1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N653
- Q8N653
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-zipper-like transcriptional regulator 1 protein (300-545AA)
- Recombinant Human Leucine-zipper-like transcriptional regulator 1 protein (300-545AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together