Anti-MOGAT1 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MOGAT1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-805
- 77720-806
- Antigen Symbol:
- MOGAT1
- MOGAT1
- Antigen Synonyms:
- mDC2|Diacylglycerol O-acyltransferase candidate 2|Acyl-CoA:monoacylglycerol acyltransferase 1|DGAT2L|Diacylglycerol acyltransferase 2 like|DGAT2L1|MOGAT1|Monoacylglycerol O acyltransferase 1|2-acylglycerol O-acyltransferase 1|hDC2|Acyl CoA:monoacylglycerol acyltransferase 1|1110064N14Rik|MOGT1_HUMAN|MGAT1|Monoacylglycerol O-acyltransferase 1|MGC118035|Diacylglycerol acyltransferase 2-like protein 1|Diacylglycerol O acyltransferase 2 like 1|MOGT1|OTTMUSP00000025181|0610030A14Rik
- mDC2|Diacylglycerol O-acyltransferase candidate 2|Acyl-CoA:monoacylglycerol acyltransferase 1|DGAT2L|Diacylglycerol acyltransferase 2 like|DGAT2L1|MOGAT1|Monoacylglycerol O acyltransferase 1|2-acylglycerol O-acyltransferase 1|hDC2|Acyl CoA:monoacylglycerol acyltransferase 1|1110064N14Rik|MOGT1_HUMAN|MGAT1|Monoacylglycerol O-acyltransferase 1|MGC118035|Diacylglycerol acyltransferase 2-like protein 1|Diacylglycerol O acyltransferase 2 like 1|MOGT1|OTTMUSP00000025181|0610030A14Rik
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96PD6
- Q96PD6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 2-acylglycerol O-acyltransferase 1 protein (163-246AA)
- Recombinant Human 2-acylglycerol O-acyltransferase 1 protein (163-246AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together