Anti-NSMCE2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NSMCE2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-780
- 77720-781
- Antigen Symbol:
- NSMCE2
- NSMCE2
- Antigen Synonyms:
- MMS21|MMS21 homolog|Non-structural maintenance of chromosomes element 2 homolog|methyl methanesulfonate sensitivity gene 21|hMMS21|non-SMC element 2|Nsmce2|NSE2_HUMAN|Non-SMC element 2 homolog|hNSE2|NSE2|C8orf36|E3 SUMO-protein ligase NSE2
- MMS21|MMS21 homolog|Non-structural maintenance of chromosomes element 2 homolog|methyl methanesulfonate sensitivity gene 21|hMMS21|non-SMC element 2|Nsmce2|NSE2_HUMAN|Non-SMC element 2 homolog|hNSE2|NSE2|C8orf36|E3 SUMO-protein ligase NSE2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96MF7
- Q96MF7
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human E3 SUMO-protein ligase NSE2 protein (1-247AA)
- Recombinant Human E3 SUMO-protein ligase NSE2 protein (1-247AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together