Anti-LRR1 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRR1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-756
- 77720-757
- Antigen Symbol:
- LRR1
- LRR1
- Antigen Synonyms:
- 4 1BBlrr|Leucine-rich repeat protein 1|LRR1|LRR-1|LRR repeat protein 1|4-1BBlrr|LRR-repeat protein 1|LRR 1|4 1BB mediated signaling molecule|4-1BB-mediated-signaling molecule|PPIL5|MGC20689|Peptidylprolyl isomerase-like 5|peptidylprolyl isomerase (cyclophilin) like 5|Cyclophilin like 5|LLR1_HUMAN|Peptidylprolyl isomerase like 5
- 4 1BBlrr|Leucine-rich repeat protein 1|LRR1|LRR-1|LRR repeat protein 1|4-1BBlrr|LRR-repeat protein 1|LRR 1|4 1BB mediated signaling molecule|4-1BB-mediated-signaling molecule|PPIL5|MGC20689|Peptidylprolyl isomerase-like 5|peptidylprolyl isomerase (cyclophilin) like 5|Cyclophilin like 5|LLR1_HUMAN|Peptidylprolyl isomerase like 5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96L50
- Q96L50
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Leucine-rich repeat protein 1 protein (18-159AA)
- Recombinant Human Leucine-rich repeat protein 1 protein (18-159AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together