Anti-ELMO2 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ELMO2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-722
- 77720-723
- Antigen Symbol:
- ELMO2
- ELMO2
- Antigen Synonyms:
- ELMO-2|hCED 12A|ELMO2|Protein ced-12 homolog A|ced-12 homolog 2|hCED12A|CED12 homolog A|Engulfment and cell motility gene 2|KIAA1834|ELMO 2|Ced 12 homolog 2|Protein ced 12 homolog A|OTTHUMP00000031762|CED 12A|C. elegans)|ELMO2_HUMAN|hCED-12A|C. elegans|Engulfment and cell motility protein 2|CED12A|Engulfment and cell motility 2 (ced 12 homolog|2|CED 12|CED 12 homolog A|homolog of|Engulfment and cell motility 2|PH domain protein CED12A|CED12|CED-12|FLJ11656
- ELMO-2|hCED 12A|ELMO2|Protein ced-12 homolog A|ced-12 homolog 2|hCED12A|CED12 homolog A|Engulfment and cell motility gene 2|KIAA1834|ELMO 2|Ced 12 homolog 2|Protein ced 12 homolog A|OTTHUMP00000031762|CED 12A|C. elegans)|ELMO2_HUMAN|hCED-12A|C. elegans|Engulfment and cell motility protein 2|CED12A|Engulfment and cell motility 2 (ced 12 homolog|2|CED 12|CED 12 homolog A|homolog of|Engulfment and cell motility 2|PH domain protein CED12A|CED12|CED-12|FLJ11656
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JJ3
- Q96JJ3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Engulfment and cell motility protein 2 protein (304-371AA)
- Recombinant Human Engulfment and cell motility protein 2 protein (304-371AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together