Anti-ZMYND19 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: ZMYND19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77720-554
- 77720-555
- Antigen Symbol:
- ZMYND19
- ZMYND19
- Antigen Synonyms:
- RP11 48C7.4|Melanin concentrating hormone receptor 1 interacting zinc finger protein|MIZIP|MCHR1 interacting zinc finger protein|Zinc finger MYND domain-containing protein 19|Zinc finger MYND domain containing 19|ZMY19_HUMAN|zinc finger MYND type containing 19|ZMYND19|Melanin-concentrating hormone receptor 1-interacting zinc finger protein|MCH-R1-interacting zinc finger protein
- RP11 48C7.4|Melanin concentrating hormone receptor 1 interacting zinc finger protein|MIZIP|MCHR1 interacting zinc finger protein|Zinc finger MYND domain-containing protein 19|Zinc finger MYND domain containing 19|ZMY19_HUMAN|zinc finger MYND type containing 19|ZMYND19|Melanin-concentrating hormone receptor 1-interacting zinc finger protein|MCH-R1-interacting zinc finger protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96E35
- Q96E35
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc finger MYND domain-containing protein 19 protein (1-227AA)
- Recombinant Human Zinc finger MYND domain-containing protein 19 protein (1-227AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together