Anti-NT5C3A Rabbit Polyclonal Antibody
77705-401
New Product
- Antigen Symbol:NT5C3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9H0P0
- Antigen Synonyms:PSN1|Uridine 5' monophosphate hydrolase 1|Pyrimidine 5''-nucleotidase 1|p36|P5''N-1|Nt5c3a|P5N 1|UMPH1|PN-I|UMPH|P5'N1|Cytosolic 5''-nucleotidase III|cNIII|Cytosolic 5''-nucleotidase 3|P5'N 1|P5N-1|Uridine 5''-monophosphate hydrolase 1|5NT3A_HUMAN|NT5C3|cN III|cN-III|PNI|Cytosolic 5''-nucleotidase 3A|Pyrimidine 5' nucleotidase 1|PN I|P5N1|Cytosolic 5' nucleotidase III|Cytosolic 5' nucleotidase 3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human NT5C3
- Purification:Antigen affinity purified
- Cat. No.:77705-401
- Packaging:Vial
Type:
Antigen: NT5C3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse