Anti-RTL6 Rabbit Polyclonal Antibody
77715-512
New Product
- Antigen Symbol:RTL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q6ICC9
- Antigen Synonyms:Leucine zipper protein down-regulated in cancer cells-like|Protein LDOC1L|LDOCL_HUMAN|Leucine zipper down regulated in cancer 1 like|Mart6|Mammalian retrotransposon-derived protein 6|Mammalian retrotransposon derived protein 6|DKFZp761O17121|Mar6|LDOC1L|dJ1033E15.2
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human LDOCL
- Purification:Antigen affinity purified
- Cat. No.:77715-512
- Packaging:Vial
WB:1:500-1:3000
Type:
Antigen: RTL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse