Anti-SRSF4 Rabbit Polyclonal Antibody
77712-114
New Product
- Antigen Symbol:SRSF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µl
- Form:Liquid
- Gene ID:Q08170
- Antigen Synonyms:SRP001LB|Pre-mRNA-splicing factor SRP75|arginine/serine-rich 4|Splicing factor|Pre mRNA splicing factor SRP75|Serine/arginine-rich splicing factor 4|SRP75|Splicing factor, arginine/serine rich 4|SRSF4_HUMAN|Srsf4|Splicing factor, arginine/serine rich, 75-KD
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human SFRS4
- Purification:Antigen affinity purified
- Cat. No.:77712-114
- Packaging:Vial
Type:
Antigen: SRSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse