Order Entry
United States
ContactUsLinkComponent
Anti-CYP2J2 Rabbit Polyclonal Antibody
Anti-CYP2J2 Rabbit Polyclonal Antibody
Catalog # 77711-265
Supplier:  AFG Bioscience
New Product
undefined
Anti-CYP2J2 Rabbit Polyclonal Antibody
Catalog # 77711-265
Supplier:  AFG Bioscience
Supplier Number:  A46476-100UL
New Product

Specifications

  • Antigen Symbol:
    CYP2J2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µl
  • Form:
    Liquid
  • Gene ID:
    P51589
  • Antigen Synonyms:
    Microsomal monooxygenase|Cytochrome P450 subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2|Cytochrome P450 subfamily IIJ polypeptide 2|Flavoprotein linked monooxygenase|Arachidonic acid epoxygenase|CYPIIJ2|CP2J2_HUMAN|Cytochrome P450 2J2|OTTHUMP00000010550|CYP2J2|Cytochrome P450 family 2 subfamily J polypeptide 2|CPJ2
  • Storage Buffer:
    PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Synthesized peptide derived from inetrnal of Human Cytochrome P450 2J2
  • Purification:
    Antigen affinity purified
  • Cat. No.:
    77711-265
  • Packaging:
    Vial

Specifications

About this item

WB:1:500-1:3000, IHC:1:50-1:100, IF:1:100-1:500

Type:
Antigen: CYP2J2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human