Anti-CACNA2D1 Rabbit Polyclonal Antibody
Catalog # 77687-295
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Symbol:CACNA2D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P54289
- Antigen Synonyms:Voltage gated calcium channel subunit alpha 2/delta 1|Voltage-dependent calcium channel subunit delta-1|MHS3|L type calcium channel subunit alpha 2|CACNL2A|MHS 3|CA2D1_HUMAN|Voltage dependent calcium channel subunit alpha 2/delta 1|CCHL2A|Cacna2d1|CACN A2|CACNA2|Calcium channel voltage dependent alpha 2/delta subunit 1|Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit|Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta|Dihydropyridine receptor alpha 2 subunit|Calcium channel L type alpha 2 polypeptide|Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Voltage-dependent calcium channel subunit alpha-2/delta-1 protein (577-717AA)
- Purification:>95%, Protein G purified
- Cat. No.:77687-295
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:500
Type:
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human