Anti-SMYD2 Rabbit Polyclonal Antibody
77700-592
New Product
- Antigen Symbol:SMYD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NRG4
- Antigen Synonyms:SMYD2_HUMAN|N-lysine methyltransferase SMYD2|SET and MYND domain containing protein 2|Smyd2|HSKM-B|HSKMB|N lysine methyltransferase SMYD2|KMT3C|Zinc finger MYND domain containing 14|SET and MYND domain-containing protein 2|HSKM B|MGC119305|Lysine N-methyltransferase 3C|Histone methyltransferase SMYD2|SET and MYND domain containing 2|ZMYND14
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human SMYD2
- Purification:Antigen affinity purified
- Cat. No.:77700-592
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: SMYD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat