Anti-SMAD3 Rabbit Polyclonal Antibody
77700-586
New Product
- Antigen Symbol:SMAD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P84022
- Antigen Synonyms:mothers against decapentaplegic homolog 3|hMAD 3|JV15 2|JV15-2|SMAD|DKFZp686J10186|JV152|DKFZP586N0721|SMA and MAD related protein 3|Mad homolog JV15 2|Mothers against DPP homolog 3|SMAD3_HUMAN|LDS3|MADH 3|HST17436|SMAD 3|MAD (mothers against decapentaplegic Drosophila) homolog 3|MAD|Mad protein homolog|SMAD family member 3|hSMAD3|Mad3|hMAD-3|HSPC193|MAD homolog 3|MGC60396|Smad3|LDS1C|MADH3
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Smad3 around the non-phosphorylation site of S425
- Purification:Antigen affinity purified
- Cat. No.:77700-586
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat