Anti-SCFD1 Rabbit Polyclonal Antibody
Catalog # 77700-539
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SCFD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8WVM8
- Antigen Synonyms:Sly1p|Sec1 family domain-containing protein 1|Vesicle transport related protein|SLY1|STXBP1L2|Syntaxin binding protein 1 like 2|C14orf163|SCFD1_HUMAN|SLY1 homolog|RA410|Scfd1|Chromosome 14 open reading frame 163|Sec1 family domain containing 1|Syntaxin-binding protein 1-like 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human SCFD1
- Purification:Antigen affinity purified
- Cat. No.:77700-539
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: SCFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat