Anti-RHOBTB1/RHOBTB2 Rabbit Polyclonal Antibody
Catalog # 77700-481
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RHOBTB1/RHOBTB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O94844/Q9BYZ6
- Antigen Synonyms:p83|Rho-related BTB domain-containing protein 1|DBC2|Deleted in breast cancer 2 gene protein|KIAA0740|KIAA0717|RHOBTB1|Rho-related BTB domain-containing protein 2|RHOBTB2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human RhoBTB1/2
- Purification:Antigen affinity purified
- Cat. No.:77700-481
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:5000
Type:
Antigen: RHOBTB1/RHOBTB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse