Anti-RTL9 Rabbit Polyclonal Antibody
77700-475
New Product
- Antigen Symbol:RTL9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8NET4
- Antigen Synonyms:Retrotransposon gag domain-containing protein 1|KIAA1318|RTL9|Retrotransposon Gag-like protein 9|Tumor antigen BJ-HCC-23|RGAG1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human RGAG1
- Purification:Antigen affinity purified
- Cat. No.:77700-475
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: RTL9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse