Anti-RAP2C Rabbit Polyclonal Antibody
77700-456
New Product
- Antigen Symbol:RAP2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9Y3L5
- Antigen Synonyms:Ras-related protein Rap-2c|OTTMUSP00000019067|2010200P20Rik|MGC143331|BOS_25229|RAP2C_HUMAN|OTTHUMP00000024037|AI194294|RP23-15D12.2|Ras related protein Rap 2c|DKFZp313B211|AL022976|member of RAS oncogene family|RAP2C
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human Rap 2C
- Purification:Antigen affinity purified
- Cat. No.:77700-456
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: RAP2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse