Anti-EIF2AK2 Rabbit Polyclonal Antibody
Catalog # 77700-363
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF2AK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P19525
- Antigen Synonyms:double-stranded RNA-activated protein kinase|Serine/threonine protein kinase TIK|PPP1R83|Protein kinase RNA activated|PRKR|Interferon-induced|EIF2AK1|eIF-2A protein kinase 2|Interferon-inducible RNA-dependent protein kinase|Protein phosphatase 1 regulatory subunit 83|P1/eIF2A protein kinase|MGC126524|Eukaryotic translation initiation factor 2-alpha kinase 2|Double stranded RNA activated protein kinase|Interferon inducible RNA dependent protein kinase|Protein kinase RNA-activated|EIF2AK2|P1/eIF-2A protein kinase|PKR|HGNC:9437|E2AK2_HUMAN|Tyrosine protein kinase EIF2AK2|Interferon induced double stranded RNA activated protein kinase|Protein kinase interferon inducible double stranded RNA dependent|Interferon inducible elF2 alpha kinase|p68 kinase|Eukaryotic translation initiation factor 2 alpha kinase 2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human PKR around the non-phosphorylation site of T446
- Purification:Antigen affinity purified
- Cat. No.:77700-363
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: EIF2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human