Anti-SLC22A18 Rabbit Polyclonal Antibody
Catalog # 77700-230
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SLC22A18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q96BI1
- Antigen Synonyms:ORCTL-2|Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene A protein|HET|SLC22A18|Tumor-suppressing STF cDNA 5 protein|ITM|Efflux transporter-like protein|Imprinted multi-membrane-spanning polyspecific transporter-related protein 1|BWR1A|ORCTL2|Organic cation transporter-like protein 2|p45-BWR1A|Solute carrier family 22 member 1-like|p45-Beckwith-Wiedemann region 1 A|BWSCR1A|SLC22A1L|TSSC5|IMPT1|Solute carrier family 22 member 18|Tumor-suppressing subchromosomal transferable fragment candidate gene 5 protein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human ORCTL2
- Purification:Antigen affinity purified
- Cat. No.:77700-230
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: SLC22A18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human