Anti-MRPL50 Rabbit Polyclonal Antibody
Catalog # 77699-997
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRPL50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q8N5N7
- Antigen Synonyms:MRPL50|mitochondrial|39S ribosomal protein L50|Mitochondrial large ribosomal subunit protein mL50|L50mt|MRP-L50
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human MRP-L50
- Purification:Antigen affinity purified
- Cat. No.:77699-997
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: MRPL50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human