Anti-MRGPRG Rabbit Polyclonal Antibody
Catalog # 77699-976
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRGPRG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q86SM5
- Antigen Synonyms:Mas-related G-protein coupled receptor member G|GPR169|MRGG|MRGPRG|G-protein coupled receptor 169
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human MRGG
- Purification:Antigen affinity purified
- Cat. No.:77699-976
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:40000
Type:
Antigen: MRGPRG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human