Anti-MMP11 Rabbit Polyclonal Antibody
Catalog # 77699-957
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MMP11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P24347
- Antigen Synonyms:Mmp11|Matrix metalloproteinase-11|SL3|ST3|Matrix Metalloproteinase 11|Stromelysin-3|STMY3|SL 3|SL-3|MMP-11|Stromelysin 3|Stromelysin III|MMP11_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human MMP-11
- Purification:Antigen affinity purified
- Cat. No.:77699-957
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: MMP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat