Anti-MAGEC2 Rabbit Polyclonal Antibody
Catalog # 77699-898
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAGEC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q9UBF1
- Antigen Synonyms:Cancer testis antigen 10|Cancer/testis antigen 10|MAGE-E1 antigen|CT10|MGC13377|Melanoma antigen family E 1 cancer/testis specific|MAGEC2|Melanoma-associated antigen C2|MAGE C2 antigen|MAGE family member C2|Melanoma antigen family C 2|Hepatocellular carcinoma-associated antigen 587|Melanoma associated antigen C2|MAGE-C2 antigen|Melanoma antigen family C2|MAGE E1 antigen|HCA587|Hepatocellular carcinoma associated antigen 587|Hepatocellular cancer antigen 587|MAGC2_HUMAN
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human MAGE-C2
- Purification:Antigen affinity purified
- Cat. No.:77699-898
- Packaging:Vial
Specifications
About this item
Type:
Antigen: MAGEC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human