Anti-AGPAT4 Rabbit Polyclonal Antibody
Catalog # 77699-879
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:AGPAT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q9NRZ5
- Antigen Synonyms:1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase|1-acylglycerol-3-phosphate O-acyltransferase 4|1-AGP acyltransferase 4|1 AGP acyltransferase 4|LPAAT delta|RP3 473J16.2|1 acylglycerol 3 phosphate O acyltransferase 4|delta)|Lysophosphatidic acid acyltransferase-delta (LPAAT-delta)|dJ473J16.2|1-AGPAT 4|Lysophosphatidic acid acyltransferase delta|LPAAT-delta|1-acyl-sn-glycerol-3-phosphate acyltransferase delta|1 acyl sn glycerol 3 phosphate acyltransferase delta|PLCD_HUMAN|AGPAT4|1 AGPAT4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human LPAAT-δ
- Purification:Antigen affinity purified
- Cat. No.:77699-879
- Packaging:Vial
Specifications
About this item
Type:
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat