Anti-IRF4 Rabbit Polyclonal Antibody
Catalog # 77699-806
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IRF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q15306
- Antigen Synonyms:IRF4_HUMAN|Lymphocyte specific IRF|MUM1|MUM 1|Transcriptional activator PIP|NF EM5|Lymphocyte specific interferon regulatory factor|IRF-4|Sfpi1/PU.1 interaction partner|PU.1 interaction partner|IRF 4|LSIRF|Interferon regulatory factor 4|NFEM5|Irf4|Multiple myeloma oncogene 1|NF-EM5|Lymphocyte-specific interferon regulatory factor
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human IRF-4
- Purification:Antigen affinity purified
- Cat. No.:77699-806
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IRF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse