Anti-HOXB4 Rabbit Polyclonal Antibody
Catalog # 77699-723
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HOXB4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P17483
- Antigen Synonyms:HOX B4|Hox2F|Homeobox protein Hox-2F|HOXB 4|Hox 2F|Homeo box 2F|Homeobox protein Hox-B4|Homeobox B4|Homeobox protein Hox B4|HOX 2|Hox2.6|HOX2|Homeobox 2F|Homeo box B4|Hox 2.6|hoxb4|HXB4_HUMAN|Homeobox protein HoxB4|Homeobox protein Hox-2.6
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human HoxB4
- Purification:Antigen affinity purified
- Cat. No.:77699-723
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: HOXB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse