Order Entry
United States
ContactUsLinkComponent
Nitric acid 5% in aqueous solution matrix blank, VHG
Corrosive
Irritant
Nitric acid 5% in aqueous solution matrix blank, VHG
Catalog # 77697-528
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Corrosive
Irritant
Nitric acid 5% in aqueous solution matrix blank, VHG
Catalog # 77697-528
Supplier:  LGC Standards
Supplier Number:  VHG-HNO3-BLK-500
New Product
CAS Number:  

Specifications

  • Size:
    500 ml
  • Cat. No.:
    77697-528

Specifications

About this item

 :
 :63.01
UN:3264
ADR:8,III
 :GHS05
 :GHS07
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349