Antimony AA Standard: Sb @ 1000 µg/mL in 30% HCl, VHG, LGC Standards
Catalog # 77693-016
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:30% HCl
- Description:Antimony AA Standard: Sb @ 1000 µg/mL in 30% HCl
- Size:100 ml
- Cat. No.:77693-016