Strontium AA Standard: Sr @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77695-315
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:5% HNO₃
- Description:Strontium AA Standard: Sr @ 1000 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77695-315