Continuing Calibration Verification Standard 2: Al, As, Cr, Sb, Se, V @ 500; Ca, K, Mg, Na @ 5000 µg/mL in 20% HCl, VHG, LGC Standards
77693-053
New Product
:
- Matrix:20% HCl
- Description:Continuing Calibration Verification Standard 2: Al, As, Cr, Sb, Se, V @ 500; Ca, K, Mg, Na @ 5000 µg/mL in 20% HCl
- Size:100 ml
- Cat. No.:77693-053