Order Entry
United States
ContactUsLinkComponent
 
Bromide (from KBr) Standard: Br- @ 10000 µg/mL in H2O, VHG, LGC Standards
Catalog # 77693-105
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Bromide (from KBr) Standard: Br- @ 10000 µg/mL in H2O, VHG, LGC Standards
Catalog # 77693-105
Supplier:  LGC Standards
Supplier Number:  VHG-I1PBR-500
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    H₂O
  • Description:
    Bromide (from KBr) Standard: Br- @ 10000 µg/mL in H2O
  • Size:
    500 ml
  • Cat. No.:
    77693-105

Specifications

About this item