CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 µg/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77693-119
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:5% HNO₃
- Description:CLP Instrument Calibration Standard 1: Ag @ 200; Be @ 400; Cd @ 500; Ba, Co, Cu, Fe, Mn, Ni, Pb, Tl, Zn @ 1000 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77693-119