CLP Instrument Calibration Standard 2: 1000 µg/mL of Al, As, Cr, Sb, Se, V; 10000 µg/mL of Ca, K, Mg, Na in 20% HCl, VHG, LGC Standards
Catalog # 77693-122
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:20% HCl
- Description:CLP Instrument Calibration Standard 2: Al, As, Cr, Sb, Se, V @ 1000; Ca, K, Mg, Na @ 10000 µg/mL in 20% HCl
- Size:100 ml
- Cat. No.:77693-122