Chromate Standard: CrO4(2-) @ 1000 µg/mL in H2O, VHG, LGC Standards
Catalog # 77693-127
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg/mL
- Matrix:H₂O
- Description:Chromate Standard: CrO4(-2) @ 1000 µg/mL in H2O
- Size:100 ml
- Cat. No.:77693-127