Order Entry
United States
ContactUsLinkComponent
Multi-Element 22 Components Calibration Standard for 6020 CLP-M: 10 µg/mL Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 2% HNO3,tr. Tartaric Acid,VHG,LGC Standards
Multi-Element 22 Components Calibration Standard for 6020 CLP-M: 10 µg/mL Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 2% HNO3,tr. Tartaric Acid,VHG,LGC Standards
Catalog # 77693-154
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Element 22 Components Calibration Standard for 6020 CLP-M: 10 µg/mL Ag,Al,As,Ba,Be,Ca,Cd,Co,Cr,Cu,Fe,K,Mg,Mn,Na,Ni,Pb,Sb,Se,Tl,V,Zn in 2% HNO3,tr. Tartaric Acid,VHG,LGC Standards
Catalog # 77693-154
Supplier:  LGC Standards
Supplier Number:  VHG-LCAL6020-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Calibration Standard for 6020 CLP-M: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, Pb, Sb, Se, Tl, V, Zn @ 10 µg/mL in 2% HNO3, tr. Tartaric Acid
  • Size:
    100 ml
  • Cat. No.:
    77693-154

Specifications

About this item