Copper Standard: Cu @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77693-157
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:2% HNO₃
- Description:Copper Standard: Cu @ 10 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77693-157