Boron Isotopic Standard: B @ 100 µg/mL in H2O (Enriched in B-11), VHG, LGC Standards
Catalog # 77693-159
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:H₂O
- Description:Boron Isotopic Standard: B @ 100 µg/mL in H2O (Enriched in B-11)
- Size:50 ml
- Cat. No.:77693-159