Order Entry
United States
ContactUsLinkComponent
Multi-Element 6 Component US EPA 6020 Internal Standard Stock Solution: 10 µg/mL each of Li, Sc, Y, In, Tb, Ho, Bi in 5% HNO3, VHG, LGC Standards
Multi-Element 6 Component US EPA 6020 Internal Standard Stock Solution: 10 µg/mL each of Li, Sc, Y, In, Tb, Ho, Bi in 5% HNO3, VHG, LGC Standards
Catalog # 77695-343
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Element 6 Component US EPA 6020 Internal Standard Stock Solution: 10 µg/mL each of Li, Sc, Y, In, Tb, Ho, Bi in 5% HNO3, VHG, LGC Standards
Catalog # 77695-343
Supplier:  LGC Standards
Supplier Number:  VHG-LIS6020-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    US EPA 6020 Internal Standard Stock Solution: 6Li, Sc, Y, In, Tb, Ho, Bi @ 10 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-343

Specifications

About this item