Order Entry
United States
ContactUsLinkComponent
 
Yttrium Internal Standard: Y @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
  77695-344
New Product
 :  
undefined
Yttrium Internal Standard: Y @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
  77695-344
 :  VHG-LISAY-100
New Product
 :  

 

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Yttrium Internal Standard: Y @ 10 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-344