Tin Standard: Sn @ 10 µg/mL in 5% HCl, VHG, LGC Standards
Catalog # 77695-345
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/mL
- Matrix:5% HCl
- Description:Tin Standard: Sn @ 10 µg/mL in 5% HCl
- Size:100 ml
- Cat. No.:77695-345