Order Entry
United States
ContactUsLinkComponent
200.8 Calibration Standard B: Ag, Ba @ 10 µg/mL in 1% HNO3, VHG, LGC Standards
200.8 Calibration Standard B: Ag, Ba @ 10 µg/mL in 1% HNO3, VHG, LGC Standards
Catalog # 77693-163
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
200.8 Calibration Standard B: Ag, Ba @ 10 µg/mL in 1% HNO3, VHG, LGC Standards
Catalog # 77693-163
Supplier:  LGC Standards
Supplier Number:  VHG-LSSB-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    1% HNO₃
  • Description:
    200.8 Calibration Standard B: Ag, Ba @ 10 µg/mL in 1% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77693-163

Specifications

About this item